Learn More
Memmert Memmert Peltier Cooled Incubator IPP55Plus. 55L - 115V, Stainless Steel Construction with TwinDISPLAY digital controller.
SDP
Catalog No. 502254382
Encompass_Preferred
Memmert Cooled-Incubators - this programmable model is the 115 volt, Peltier-technology seamlessly heats and cools, with TwinDISPLAY digital controller. Interior dimensions 400/400/320-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Atmocontrol Software, Manual, Quick Start Guide, Calibration Certificate and Power Cord - energy efficient, and precise! Working temperature range 0C (at least 20 below ambient) to +70C.
This item is not returnable. View return policy
Catalog No. 50-225-4382 Supplier Memmert Supplier No. IPP55 PLUS 115V
Add to Cart
Add to Cart
Request a Quote
Memmert Cooled-Incubators - this programmable model is the 115 volt, Peltier-technology seamlessly heats and cools, with TwinDISPLAY digital controller. Interior dimensions 400/400/320-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Atmocontrol Software, Manual, Quick Start Guide, Calibration Certificate and Power Cord - energy efficient, and precise! Working temperature range 0C (at least 20 below ambient) to +70C.