Learn More
Memmert Memmert Peltier Cooled Incubator IPP110Eco Plus. 110L - 115V, Stainless Steel Construction with TwinDISPLAY digital controller.
SDP
Catalog No. 502254384
Encompass_Preferred
Memmert Cooled-Incubators - this programmable model is the 115 volt, Peltier-technology seamlessly heats and cools, with TwinDISPLAY digital controller. Interior dimensions 560/480/368-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software + Manual, Quick Start Guide, Calibration Certificate and Power Cord - energy efficient, and precise! Working temperature range 0C (at least 20 below ambient) to +70C.
This item is not returnable. View return policy
Catalog No. 50-225-4384 Supplier Memmert Supplier No. IPP110ECO PLUS 115V
Add to Cart
Add to Cart
Request a Quote
Memmert Cooled-Incubators - this programmable model is the 115 volt, Peltier-technology seamlessly heats and cools, with TwinDISPLAY digital controller. Interior dimensions 560/480/368-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software + Manual, Quick Start Guide, Calibration Certificate and Power Cord - energy efficient, and precise! Working temperature range 0C (at least 20 below ambient) to +70C.