Learn More
Sheldon Manufacturing Inc PELTIER-REFRIGERATED INCUBATOR
SDP
Catalog No. NC0981562
Encompass_Preferred
REFRIGERATED INCUBATOR, 3 CU FT, ENERGY EFFICIENT PELTIER COOLING, +15c to +40c, OUTLET, 115V. 60 Bottle capacity. Using innovative thermoelectric cooling technology, the LI6P eliminates the need for a refrigeration compressor. This means that these units use 78% less power than alternative models and reduce room air conditioning loads by 75%. They also include 75 pound capacity shelves, which eliminates sagging. These incubators meet APHA specifications for Biochemical Oxygen Demand (BOD) and include a mechanical convection system to ensure even air distribution, digital temperature set controller, over temperature limit control, and a digital temperature display. Low Temperature Incubators are ideal for testing of sewage and wastewater, as well as other low and room temperature applications. More information available at www.shellab.com
Catalog No. NC0981562 Supplier Sheldon Manufacturing Inc Supplier No. SRI3P
REFRIGERATED INCUBATOR, 3 CU FT, ENERGY EFFICIENT PELTIER COOLING, +15c to +40c, OUTLET, 115V. 60 Bottle capacity. Using innovative thermoelectric cooling technology, the LI6P eliminates the need for a refrigeration compressor. This means that these units use 78% less power than alternative models and reduce room air conditioning loads by 75%. They also include 75 pound capacity shelves, which eliminates sagging. These incubators meet APHA specifications for Biochemical Oxygen Demand (BOD) and include a mechanical convection system to ensure even air distribution, digital temperature set controller, over temperature limit control, and a digital temperature display. Low Temperature Incubators are ideal for testing of sewage and wastewater, as well as other low and room temperature applications. More information available at www.shellab.com