Learn More
Perkin Elmer US LLC Peltier Temperature Controller
Catalog No. 502104376
Encompass_Preferred
This Peltier Temperature Controller Unit is required for operation of either the 100-position (N2930035) or Double Microtiter Peltier trays (N2930037).
Catalog No. 50-210-4376 Supplier Perkin Elmer US LLC Supplier No. N2930036
This Peltier Temperature Controller Unit is required for operation of either the 100-position (N2930035) or Double Microtiter Peltier trays (N2930037).