Learn More
MSC 3M™ PELTOR™ X4A Over-the-Head Earmuffs, Hi-Viz, Headband, 33 dB, 10/Case
Catalog No. 5000684282
Encompass
These over-the-head earmuffs are designed to provide effective hearing protection in high-noise environments. Featuring a low-profile cup design, they offer improved compatibility with other personal protective equipment while maintaining a high ratio of noise reduction to earcup depth.
  • Noise reduction rating of 25–27 dB
  • Low-profile design for enhanced compatibility with other ppe
  • Slow-recovery, high-density foam for consistent performance
  • Testable with the dual-ear validation system
  • Replaceable ear cushions and foam liners available via hygiene kit hyx4
Catalog No. 50-006-84282 Supplier MSC Supplier No. 62698501
These over-the-head earmuffs are designed to provide effective hearing protection in high-noise environments. Featuring a low-profile cup design, they offer improved compatibility with other personal protective equipment while maintaining a high ratio of noise reduction to earcup depth.
  • Noise reduction rating of 25–27 dB
  • Low-profile design for enhanced compatibility with other ppe
  • Slow-recovery, high-density foam for consistent performance
  • Testable with the dual-ear validation system
  • Replaceable ear cushions and foam liners available via hygiene kit hyx4