Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ X Series Forestry Ear Muffs

Catalog No. 19153672 Shop All 3M Products
Change view
Click to view available options
Model:
X1P5E-OR
Style:
Hard Hat
Noise Reduction Rating (NRR):
21 dB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Style Noise Reduction Rating (NRR)
19-153-672 X1P5E-OR Hard Hat 21 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-153-672 Supplier 3M™ Supplier No. 7100097447

missing translation for 'validMainframeMsg'

PELTOR™ X Series Forestry Ear Muffs are electrically insulated with orange coloring to improve visibility for use in the forestry and other applicable industries.

  • Ideal for use in forestry and landscaping industries
  • Electrically insulated
  • Orange color is highly visible
  • High-performance arms to balance durability and flexibility
  • Easy click in/click out
  • Hard hat compatibility (models X1P5E and X4P5E)

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Equipment) Fits a wide range of industrial hard hats
For Use With (Application) Forestry, Landscaping, Utilities, Mining, Oil and Gas, Electrical Contractors
High Visibility Yes
Model X1P5E-OR
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 21 dB
Product Type X Series Forestry Ear Muffs
Dielectric Yes
Material Foam, Plastic, Metal
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.