Learn More
3M™ PELTOR™ EEP-100 Electronic Ear Plug
Catalog No. 19071057 Shop All 3M Products
Change view
Click to view available options
PELTOR™ Electronic Ear Plug; EEP-100
Catalog No.
19-071-057 PELTOR™ Electronic Ear Plug; EEP-100
1 options
Catalog No. 19-071-057 Supplier 3M™ Supplier No. 7100193144
Add to Cart
Add to Cart
3M™ PELTOR™ EEP-100 Electronic ear plug protects hearing and can help improve situational awareness and communications in challenging environments. Kit includes a protective charging case and an assortment of ear tips.
3M™ PELTOR™ electronic ear pieces are rechargeable via a USB jack on the charging case. The case also provides easy and convenient storage of the earpieces when not being used.
The small and lightweight design helps improve the overall comfort and user experience, and makes it compatible with most head-borne 3M Personal Safety Equipment (PPE), such as helmets and face shields.
Key Features:
  • Helps provide hearing protection
  • Level-dependent function (for situational awareness)
  • Intuitive one-button operation
  • Rechargeable
  • Durable storage and charging case (IP54)
  • Small and lightweight design
  • Compatible with most head borne 3M Personal Protective Equipment
Includes:
  • 2 x Ear pieces
  • 1 x Charging case
  • 2 x 3M™ UltraFit Communication Tips Replacement, Large
  • 2 x 3M™ UltraFit Communication Tips Replacement, Medium
  • 2 x 3M™ UltraFit Communication Tips Replacement, Small
  • 2 x 3M™ PELTOR™ Communication Tip, Triple C
  • 1 x USB cable
  • 1 x User Instructions
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Maintenance, Manufacturing, Shooting
High Visibility No
Model EEP-100
Pairs per Package 1 Pair/Pk.
Plug Material Elastomeric polymer
Plug Color Tan
Noise Reduction Rating (NRR) 23 dB (Ultrafit™), 27 dB (Triple-C)
Corded Uncorded
Plug Shape Flanged
Disposable Reusable
Metal Detectable No