Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ FLX2 Cable for CH-3 Headset With Built-In PTT Button

Catalog No. 19072310 Shop All 3M Products
Change view
Click to view available options
Model:
FLX2-101
FLX2-107-50
FLX2-111
FLX2-18
FLX2-21
FLX2-28
FLX2-32
FLX2-35
FLX2-36
FLX2-44
FLX2-63-50
FLX2-64
FLX2-65
For Use With (Equipment):
2.5 Hytera, Icom, Kenwood, Motorola
Hytera PD7
Icom 2-Pin Angled
Icom F34/44
Icom Multipin S8
Kenwood Multi-pin
Kenwood Two-Pin
Motorola APX/XPR
Motorola CP200/2-pin
Motorola HT Series
Motorola XTS
Motorola ex500/ex600
Sepura 800/900
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model For Use With (Equipment)
19-072-310 FLX2-107-50 Kenwood Multi-pin
19-072-257 FLX2-63-50 Motorola APX/XPR
19-072-284 FLX2-32 Motorola HT Series
19-072-293 FLX2-36 Kenwood Two-Pin
19-072-305 FLX2-21 Motorola CP200/2-pin
19-072-256 FLX2-28 2.5 Hytera, Icom, Kenwood, Motorola
19-072-309 FLX2-18 Motorola XTS
19-072-261 FLX2-65 Motorola ex500/ex600
19-072-267 FLX2-64 Icom F34/44
19-072-271 FLX2-44 Icom Multipin S8
19-072-279 FLX2-35 Icom 2-Pin Angled
19-072-264 FLX2-111 Hytera PD7
19-072-268 FLX2-101 Sepura 800/900
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 19-072-310 Supplier 3M™ Supplier No. 7100185797
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PELTOR™ FLX2 platform allows workers to pair protective communication CH-3 headsets with existing handheld devices using a built-in push-to-talk button.

  • FLX2 Cables for use with PELTOR™ CH-3 Headset with push to talk (PTT) button
  • Coiled polyurethane cable
  • Aramid fiber reinforced
  • Flexible and adaptable

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Kenwood Multi-pin
For Use With (Application) For Use with CH-3 Headsets with PTT Button
Model FLX2-107-50
Pairs per Package 1 Ea.
Product Type FLX2 Cable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.