Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ Hygiene Kit for FM Radio, ProTac, and LiteCom Headsets

Catalog No. 19072322 Shop All 3M Products
Change view
Click to view available options
Hygiene Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-072-322 Hygiene Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-322 Supplier 3M™ Supplier No. XH001651047

missing translation for 'validMainframeMsg'

Hygiene Kit fits a wide variety of 3M™ PELTOR™ Headsets and includes 1 pair of ear cushions, foam inserts and rings.

  • Hygiene kit includes 1 pair of replacement ear cushions, foam liners and rings
  • Use for PELTOR™, ProTac, Tactical™, XP, FM-Radio, Alert™ and LiteCom™ Headsets
  • Recommended replacement at least twice a year to ensure constant attenuation, hygiene and comfort

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Includes Ear cushions and foam inserts
For Use With (Equipment) For use on a wide variety of 3M™ PELTOR™ Headsets
Model HY79
Pairs per Package 1 Ea.
Product Type Hygiene Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.