Learn More
MSC PELTOR POLYCRB FACESHIELD
Catalog No. 5000787250
Encompass
This clear replacement window for face shields is designed for use with 3M Peltor™ Hard Hat Adapter P3EV/2 and 3M Peltor™ Cap-Mounted Earmuff, providing protection during various industrial applications.
  • Replacement window for face shields
  • Bright/uncoated
  • Made from polycarbonate
  • No lens shade
  • Window height of 6.75 in (17.15 cm)
  • Window width of 13.5 in
  • Meets ANSI Z87.1 standards
  • Thickness of 0.13 in (3.30 mm)
  • Suitable for various applications including welding, sanding, machine operations, demolition, cleaning, remediation, electrical, grinding, assembly, painting, and facility maintenance
Catalog No. 50-007-87250 Supplier MSC Supplier No. 62951876
This clear replacement window for face shields is designed for use with 3M Peltor™ Hard Hat Adapter P3EV/2 and 3M Peltor™ Cap-Mounted Earmuff, providing protection during various industrial applications.
  • Replacement window for face shields
  • Bright/uncoated
  • Made from polycarbonate
  • No lens shade
  • Window height of 6.75 in (17.15 cm)
  • Window width of 13.5 in
  • Meets ANSI Z87.1 standards
  • Thickness of 0.13 in (3.30 mm)
  • Suitable for various applications including welding, sanding, machine operations, demolition, cleaning, remediation, electrical, grinding, assembly, painting, and facility maintenance