Learn More
3M™ PELTOR™ ProTac III Headset Hearing Protector
Catalog No. 19072353 Shop All 3M Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
23 dB
Catalog No. Noise Reduction Rating (NRR)
19-072-353 23 dB
1 options
For industrial/occupational use only. Not for consumer sale or use.
Catalog No. 19-072-353 Supplier 3M™ Supplier No. 7100088423/DEL
For industrial/occupational use only. Not for consumer sale or use.
3M™ ProTac III Headset protects against hazardous and nuisance noise, while allowing environmental sounds to be reproduced by the electronic circuit, limited to 82 dB.
  • Professional-Quality Hearing Protection (26dB NRR)
  • Environmental microphones for level-dependent hearing protection and ambient listening.
  • Increases ability to communicate with nearby colleagues as well as hear warning signals, approaching vehicles and sounds of machines and processes
  • 3.5 mm listen-only stereo input for connection to external devices (e.g. cell phone, two-way radio, Ipod).
  • Voice guided menu system for ease of use and adjusting settings.
  • Auto power off, the headset will turn off after 4 hrs of non-use to save battery.
  • Low-battery warning to inform you when to change batteries
  • To reduce corrosion caused by sweat and moisture, the electronics are located in a separate compartment of the cup
  • Occupational use only.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
ANSI S3.1974 Yes
Cushion Material PVC Foil and PUR Foam
CSA Z94.2 Class A
Cushion Color Black
Size Universal
Model MT13H221P3E
Noise Reduction Rating (NRR) 23 dB
Disposable Reusable
Metal Detectable No
Headband Color Black
Dielectric No
Style Hardhat Attached
Foldable No
Show More Show Less
3M industrial and occupational products are intended, labeled, and packaged for sale to trained industrial and occupational customers for workplace use. Unless specifically stated otherwise on the applicable product packaging or literature, these products are not intended, labeled, or packaged for sale to or use by consumers (e.g., for home, personal, primary or secondary school, recreational/sporting, or other uses not described in the applicable product packaging or literature), and must be selected and used in compliance with applicable health and safety regulations and standards (e.g., U.S. OSHA, ANSI), as well as all product literature, user instructions, warnings, and other limitations, and the user must take any action required under any recall, field action, or other product use notice. Misuse of 3M industrial and occupational products may result in injury, sickness, death, or property damage. For help with product selection and use, consult your on-site safety professional, industrial hygienist, or other subject matter expert.