Learn More
3M™ PELTOR™ ProTac III Headset Hearing Protector
Catalog No. 19072175 Shop All 3M Products
Change view
Click to view available options
Includes:
Attached Hard Hat
Catalog No. Includes
19-072-175 Attached Hard Hat
1 options
Catalog No. 19-072-175 Supplier 3M™ Supplier No. 7100088423
Add to Cart
Add to Cart
Helps protect against hazardous and nuisance noise, while allowing environmental sounds to be reproduced by the electronic circuit, limited to 82dB
  • Professional-Quality Hearing Protection
  • Helps protect hearing from impulse noise and increases the ability to communicate with nearby colleagues, as well as hear warning signals, approaching vehicles, and sounds in machines and processes.
  • Environmental microphones for level-dependent hearing protection and ambient listening.
  • 3.5 mm listen-only stereo input for connection to external devices (e.g. cell phone, two-way radio, Ipod).
  • Voice guided menu system for ease of use and adjusting settings.
  • Auto power off, the headset will turn off after 4 hrs of non-use to save battery.
  • Low-battery warning to inform you when to change batteries
  • To reduce corrosion caused by sweat and moisture, the electronics are located in a separate compartment of the cup
  • Available in both in headband and hard hat attached versions (black).
  • Occupational use only.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance 23dB NRR
Height (Metric) 200 mm
Model MT13H221P3E
Width (Metric) 110 mm
Includes Attached Hard Hat