Learn More
3M™ Peltor™ Push-To-Talk Adapter
Catalog No. 191501600 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Astro Sabre Series Radios: I, II, III-R (requires Motorola Models: NTN5213C/NTN5664D)-NATO Wired
Kenwood Radios: TK220/320, 240/340, 240D/340D, Free Talk, Free Talk XL and Pro Talk
Motorola HT750 Series Radios-NATO Wired
Motorola Radios: FRS Single Jack-NATO Wired
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-150-1600 Motorola Radios: FRS Single Jack-NATO Wired
19-150-1597 Motorola HT750 Series Radios-NATO Wired
19-150-1593 Kenwood Radios: TK220/320, 240/340, 240D/340D, Free Talk, Free Talk XL and Pro Talk
19-150-1595 Astro Sabre Series Radios: I, II, III-R (requires Motorola Models: NTN5213C/NTN5664D)-NATO Wired
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-150-1600 Supplier 3M™ Supplier No. 7010315073
Push-To-Talk adapters for Motorola and Kenwood Radios - NATO Wired
Use 3M™ PELTOR™ Push-To-Talk (PTT) Adapters to optimize the placement of the PTT button for convenient activation
  • For use with 3M™ PELTOR™ COMTAC™ Protective Communication Headsets and compatible two-way radios
  • Ambidextrous design allows for easy use by either the right or left hand
  • Mounting clip can be rotated 360 degrees
  • Design allows for either belt or shoulder mounting
  • Button profile and tactile feedback suitable for gloved hands
  • Optional shroud protects PTT button from accidental activation, for instance when laying in the prone position
  • NEXUS TP-120 connection serves as a break-away connector for removal in emergency situations
  • Curly cable downlead with radio specific connector. To determine the appropriate PTT adapter for your two-way radio, please visit 3M.com/PeltorAdapters
  • Motorola Sabre Series Adaptor is designed to fit the size and shape of both right and left hands
    • Buttons recessed to reduce accidental activation
    • Clip on the backside can be twisted 360 degrees
    • TP-120 connection to the unit serves as a break-away connector for removal in emergency situations
    • 3M Catalog# FL5012-02, FL5014-02, FL5030-02, FL5035-02, FL5082-02
TRUSTED_SUSTAINABILITY
Specifications
Product Type Push-to-Talk Adaptor
For Use With (Equipment) Motorola Radios: FRS Single Jack-NATO Wired
Model FL5082-02