Learn More
3M™ PELTOR™ Tactical Ear Plug Replacement Case
Catalog No. 19164142 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
3M™ PELTOR™ TEP-100 and TEP-200 tactical ear plugs
Catalog No. For Use With (Equipment)
19-164-142 3M™ PELTOR™ TEP-100 and TEP-200 tactical ear plugs
1 options
Catalog No. 19-164-142 Supplier 3M™ Supplier No. 7100066370
Add to Cart
Add to Cart
Replacement charging station case for 3M™ PELTOR™ tactical ear plugs, TEP-100 and TEP-200.
  • Replacement charging station case to be used with tactical ear plugs TEP-100 and TEP-200
  • Powered by 3 AA alkaline batteries or USB (Micro B) port
  • Portable
  • Black
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance IP67
For Use With (Equipment) 3M™ PELTOR™ TEP-100 and TEP-200 tactical ear plugs
Model TEP-100 C
Product Type Replacement Case