Learn More
3M™ PELTOR™ UltraFit Communication Tip Replacement
Catalog No. 19164144 Shop All 3M Products
Change view
Click to view available options
Model:
370-TEPL-25
370-TEPM-25
370-TEPS-25
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size
19-164-144 370-TEPS-25 Small
19-164-145 370-TEPM-25 Medium
19-164-146 370-TEPL-25 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-164-144 Supplier 3M™ Supplier No. 7100066371
Add to Cart
Add to Cart
UltraFit Communication Replacement Tips are offered in three size variations and are designed for the 3M™ PELTOR™ TEP-100 Tactical ear plugs.
  • Provides protection while enabling user to hear in noisy environments
  • Compatible with 3M™ PELTOR™ TEP-100 Tactical Ear Plugs
  • Tri-Flange design for comfort and stability
  • Available in three sizes - small, medium or large
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Size Small
For Use With (Equipment) PELTOR™ TEP-100 tactical ear plugs
Model 370-TEPS-25
Plug Color Brown
Plug Shape Flanged
Product Type Replacement Parts and Accessories