Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ WS™ ALERT™ XP Headset

Catalog No. 19040630 Shop All 3M Products
Change view
Click to view available options
Model:
MRX21A2WS6-NA
MRX21P3E2WS6-NA
Style:
Hard Hat
Over-the-head
Noise Reduction Rating (NRR):
23 dB
24 dB
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Style Noise Reduction Rating (NRR)
19-040-630 MRX21P3E2WS6-NA Hard Hat 24 dB
19-040-626 MRX21A2WS6-NA Over-the-head 23 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-040-630 Supplier 3M™ Supplier No. 7100147152

missing translation for 'validMainframeMsg'

PELTOR™ WS™ ALERT™ XP Headset protects users against hazardous noise, but also makes it easy to make phone calls with both hands free in noisy environments.

  • High attenuating hearing protector available in two styles - stand-alone headband or safety helmet attachment
  • Bluetooth™ technology for connection to one external device for hands-free talk and streaming
  • Noise canceling boom microphone for clear speech transmission
  • Level-dependent function for ambient listening to help improve situational awareness
  • Auto-pairing to quickly connect to mobile devices
  • Built-in FM-Radio with radio volume limited to 82dB
  • Voice guided menu system
  • Digital station search with ability to store up to 4 favorite stations
  • Auto power will turn off after 4 hrs of non-use to save battery
  • Low-battery warning at low battery level

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Equipment) Wide variety of hard hat styles
For Use With (Application) Maintenance and Manufacturing
Model MRX21P3E2WS6-NA
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 24 dB
Product Type Protective Communication
Dielectric No
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.