Learn More
3M™ PELTOR™ WS™ LiteCom PRO III Headset
Catalog No. 19072177 Shop All 3M Products
Change view
Click to view available options
Includes:
Hard Hat Attached with Bluetooth™
Hard Hat Attached with Bluetooth™ and a jack to connect to external devices
Headband with Bluetooth™
Headband with Bluetooth™ and a jack to connect to external devices
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-072-177 Headband with Bluetooth™ and a jack to connect to external devices
19-072-178 Hard Hat Attached with Bluetooth™ and a jack to connect to external devices
19-072-179 Headband with Bluetooth™
19-072-180 Hard Hat Attached with Bluetooth™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-072-177 Supplier 3M™ Supplier No. 7100099790
Add to Cart
Add to Cart
Request a Quote
A hearing protector with built-in analog and digital programmable two-way communication radio with level-dependent hearing protection.
  • Built-in programmable two-way communication radio allows for clear, hands-free communication in high levels of noise without cumbersome cables or cords.
  • The headset can be programmed to use up to 70 unique two-way radio channels using analog and/or DMR digital modulation.
  • Simultaneous, seamless connection to multiple external devices using Bluetooth™ technology.
  • Noise attenuating ear cups help provide hearing protection in environments with potentially hazardous noise.
  • Environmental listening with level dependent hearing protection for face-to-face communication and situational awareness.
  • Noise-cancelling waterproof speech microphone with voice operated transmission (VOX) for hands-free operation; helps provide clear communication in noisy and demanding environments.
  • User-friendly controls with voice-guided menu.
  • Built tough for the working professional.
  • Headset is available in headband, neckband and hard hat attached configurations.
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 224 mm
Model MT73H7A4D10NA
Width (Metric) 155 mm
Includes Headband with Bluetooth™ and a jack to connect to external devices