Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ X5 Series Ear Muffs

Catalog No. 17000640 Shop All 3M Products
Change view
Click to view available options
Model:
X5A
X5P3E
Dielectric:
No
Yes
Style:
Hard Hat
Over-the-head
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Dielectric Style
17-000-640 X5P3E No Hard Hat
17-000-635 X5A Yes Over-the-head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-000-640 Supplier 3M™ Supplier No. 7000104079

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PELTOR™ X5 Series Ear Muffs feature a combination of specially-formulated foam plus an innovative spacer and cup design to make it ideal for very high level noise situations.

  • Unparalleled attenuation without the need for double protection
  • Offered in over-the-head or hard hat-attached styles
  • Twin headband design helps reduce heat buildup with good fit and balance
  • Combination of new technologies allows for more room inside the ear cup for a more comfortable fit
  • Ear cup pivot points tilt for optimum comfort and efficiency
  • Dielectric (models X5A and X5P5E only)
  • Compatibility with hard hats (models X5P3E and X5P5E)

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black, Gray
ANSI S3.1974 Yes
Cushion Material Foam
CSA Z94.2 Class AL
Cushion Color Black
For Use With (Equipment) Fits a wide variety of industrial hard hats
For Use With (Application) Commercial Building, Construction, Heavy Infrastructure, Industrial Maintenance
Model X5P3E
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 31 dB
Product Type X5 Series
Dielectric No
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.