Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ Earmuffs, Yellow

Catalog No. 19153674 Shop All 3M Products
Change view
Click to view available options
Model:
X2A
X2P3E
X2P5E
Dielectric:
No
Yes
Style:
Hard Hat
Over-the-head
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Dielectric Style
19-153-674 X2P5E Yes Hard Hat
17-000-632 X2A Yes Over-the-head
17-000-637 X2P3E No Hard Hat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-153-674 Supplier 3M™ Supplier No. 7100097448

missing translation for 'validMainframeMsg'

3M™ PELTOR™ Earmuffs are our innovative hearing protectors with a modern slim cup design.

  • Reduces noise levels
  • Colour-coding makes it easy to identify the level of protection being worn
  • Slim cups offer improved compatibility with other safety equipment
  • Wide soft ear cushions help workers stay comfortable
  • Designed to optimise weight distribution and balance for outstanding comfort
  • ABS plastic cups are rigid, high strength and impact resistant
  • Thermoplastic polyurethane (TPU) soft outer cup coating protects from abrasion and bumps

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black, Yellow
ANSI S3.1974 Yes
Cushion Material Foam
CSA Z94.2 Class A
Cushion Color Black
For Use With (Equipment) Fits a Wide Range of Industrial Safety Hard Hats
For Use With (Application) Grinding, Machine Operations, Welding
Model X2P5E
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 24 dB
Product Type X2 Series
Dielectric Yes
Material Foam, Plastic, Metal
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.