Learn More
Selleck Chemical LLC pemafibrate-5mg
SDP
Supplier:  Selleck Chemical LLC E28765MG
Encompass_Preferred
Pemafibrate(K-877) is a selective agonist of peroxisome proliferator-activated receptor (PPAR) α with an IC50 of 10 µM.
Catalog No. 50-238-9642