Learn More
Medchemexpress LLC Pemafibrate-d4 ((R)-K-13675-d4) | 2924193-31-5 | 99.8% | 494.57 | 5 MG
SDP
Catalog No. 5000387428
Encompass_Preferred
Pemafibrate-d4 is a deuterated form of Pemafibrate, a highly selective PPARα agonist with an EC50 of 1 nM. This compound is valuable as a tracer and internal standard for quantitative analysis. Its deuteration can influence the pharmacokinetic and metabolic profiles of drugs, making it useful in drug development.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Highly selective PPARα agonist
  • Influences pharmacokinetic and metabolic profiles
Catalog No. 50-003-87428 Supplier Medchemexpress LLC Supplier No. HY17618S5MG
Pemafibrate-d4 is a deuterated form of Pemafibrate, a highly selective PPARα agonist with an EC50 of 1 nM. This compound is valuable as a tracer and internal standard for quantitative analysis. Its deuteration can influence the pharmacokinetic and metabolic profiles of drugs, making it useful in drug development.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Highly selective PPARα agonist
  • Influences pharmacokinetic and metabolic profiles