Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the estimation of Pembrolizumab in serum and plasma. Can be used for generics and biosimilar mAb PK studies. Assay Range: 10 - 640 ng/ml; Sensitivity: 8.25 ng/ml.
Supplier:  KRISHGEN BIOSYSTEMS KBI1084
Encompass_Preferred
Pack Size: 1 x 96 wells. Background: Pembrolizumab, sold under the brand name Keytruda, is a humanized antibody used in cancer immunotherapy that treats melanoma, lung cancer, head and neck cancer, Hodgkin lymphoma, stomach cancer, and cervical cancer. It is an immune checkpoint inhibitor against PD-L1, inhibiting lymphocytes' PD-1 receptors, blocking the ligands that would deactivate it and prevent an immune response. It is on the World Health Organization's List of Essential Medicines. This mAb-based Pembrolizumab ELISA accurately quantifies pembrolizumab and biosimilars in human samples. If you are looking to use animal samples, please contact us. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <15%.
Catalog No. NC2747083