Learn More
Genscript Corporation Antibody Kit Other Products Immunodetection 1/EA
Supplier:  Genscript Corporation L00968
Encompass
GenScripts Pembrolizumab Pharmacokinetic ELISA Kit had been comprehensively developed and validated for quantitative measurement of Pembrolizumab in cynomolgus monkey serum and plasma based on the ICH M10 and the FDA bioanalytical method validation guidance for industry.
Catalog No. 50-252-9977