Learn More
Medchemexpress LLC HY-10820 100mg Medchemexpress, Pemetrexed LY231514 CAS:137281-23-3 Purity:98%
SDP
Catalog No. 501637378
Encompass_Preferred
Medchemexpress,HY-10820 100mg Pemetrexed LY231514 CAS:137281-23-3 Formula:C20H21N5O6 Pemetrexed is a novel antifolate, the Ki values of the pentaglutamate of LY231514 are 1.3, 7.2, and 65 nM for inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT), respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7378 Supplier Medchemexpress LLC Supplier No. HY10820100MG
Medchemexpress,HY-10820 100mg Pemetrexed LY231514 CAS:137281-23-3 Formula:C20H21N5O6 Pemetrexed is a novel antifolate, the Ki values of the pentaglutamate of LY231514 are 1.3, 7.2, and 65 nM for inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT), respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.