Learn More
Selleck Chemical LLC Pemetrexed 200mg 150399-23-8 LY-231514
SDP
Supplier:  Selleck Chemical LLC S1135200MG
Encompass_Preferred
Pemetrexed (LY-231514) is a novel antifolate and antimetabolite for TS, DHFR and GARFT with Ki of 1.3 nM, 7.2 nM and 65 nM in cell-free assays, respectively. Pemetrexed induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-673-0