Learn More
Selleck Chemical LLC Pemetrexed disodium S1135-1g
SDP
Catalog No. 503108208
Encompass_Preferred
Pemetrexed disodium is a novel antifolate and antimetabolite for TS DHFR and GARFT with Ki of 1 3 nM 7 2 nM and 65 nM in cell-free assays respectively Pemetrexed induces autophagy and apoptosis
Catalog No. 50-310-8208 Supplier Selleck Chemical LLC Supplier No. S11351G
Pemetrexed disodium is a novel antifolate and antimetabolite for TS DHFR and GARFT with Ki of 1 3 nM 7 2 nM and 65 nM in cell-free assays respectively Pemetrexed induces autophagy and apoptosis