Learn More
Medchemexpress LLC Pemetrexed disodium | 150399-23-8 | MFCD07779402 | 99.8% | 471.37 g·mol⁻¹ | C20H19N5Na2O6 | 1 ML
SDP
Catalog No. 5000242328
Encompass_Preferred
Pemetrexed disodium is the disodium salt of pemetrexed, an antifolate antimetabolite used in research. Supplied as a ready-to-use 10 mM solution (1 mL) in water, this reagent is intended for biochemical and cell-based assays and analytical applications.
Catalog No. 50-002-42328 Supplier Medchemexpress LLC Supplier No. HY10820A10MM 1ML
Pemetrexed disodium is the disodium salt of pemetrexed, an antifolate antimetabolite used in research. Supplied as a ready-to-use 10 mM solution (1 mL) in water, this reagent is intended for biochemical and cell-based assays and analytical applications.