Learn More
TARGETMOL CHEMICALS INC Pemetrexed disodium 500MG
SDP
Catalog No. 502880683
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Pemetrexed disodium (LY-231514) is a parenterally administered folate antagonist and antineoplastic agent, used in the treatment of non-small cell lung cancer and malignant mesothelioma. Pemetrexed disodium therapy has been associated with moderate rates of serum enzyme elevations during therapy, but has not been convincingly linked to instances of acute, clinically apparent liver injury. Purity 99.92%
Catalog No. 50-288-0683 Supplier TARGETMOL CHEMICALS INC Supplier No. T0189L500MG
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Pemetrexed disodium (LY-231514) is a parenterally administered folate antagonist and antineoplastic agent, used in the treatment of non-small cell lung cancer and malignant mesothelioma. Pemetrexed disodium therapy has been associated with moderate rates of serum enzyme elevations during therapy, but has not been convincingly linked to instances of acute, clinically apparent liver injury. Purity 99.92%