Learn More
Medchemexpress LLC Pemetrexed disodium hemipenta hydrate | 357166-30-4 | 99.78% | 1 ML
SDP
Catalog No. 5000350584
Encompass_Preferred
Pemetrexed disodium hemipenta hydrate is a CNS-penetrant antifolate. Its pentaglutamate form inhibits thymidylate synthase (TS) with a Ki value of 1.3 nM, dihydrofolate reductase (DHFR) with a Ki value of 7.2 nM, and glycinamide ribonucleotide formyltransferase (GARFT) with a Ki value of 65 nM. Its antitumor activity may stem from simultaneous and multiple inhibition of several key folate-requiring enzymes via its polyglutamated metabolites. It is a novel classical antifolate.
  • CNS-penetrant antifolate
  • Inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT)
  • Potential antitumor activity through polyglutamated metabolites
  • Soluble at 10 mM in water
Catalog No. 50-003-50584 Supplier Medchemexpress LLC Supplier No. HY1378110MM/1ML
Pemetrexed disodium hemipenta hydrate is a CNS-penetrant antifolate. Its pentaglutamate form inhibits thymidylate synthase (TS) with a Ki value of 1.3 nM, dihydrofolate reductase (DHFR) with a Ki value of 7.2 nM, and glycinamide ribonucleotide formyltransferase (GARFT) with a Ki value of 65 nM. Its antitumor activity may stem from simultaneous and multiple inhibition of several key folate-requiring enzymes via its polyglutamated metabolites. It is a novel classical antifolate.
  • CNS-penetrant antifolate
  • Inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT)
  • Potential antitumor activity through polyglutamated metabolites
  • Soluble at 10 mM in water