Learn More
Medchemexpress LLC Pemetrexed disodium hemipenta hydrate | 357166-30-4 | 99.78% | 516.41 | 100 MG
SDP
Catalog No. 5000350660
Encompass_Preferred
Pemetrexed disodium hemipenta hydrate is a CNS-penetrant antifolate. Its pentaglutamate form inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT). This product is intended for research use only.
  • CNS-penetrant antifolate
  • Inhibits thymidylate synthase (TS)
  • Inhibits dihydrofolate reductase (DHFR)
  • Inhibits glycinamide ribonucleotide formyltransferase (GARFT)
  • Antitumor activity through inhibition of multiple key folate-requiring enzymes
  • Substrate for the enzyme FPGS
  • Soluble in water
Catalog No. 50-003-50660 Supplier Medchemexpress LLC Supplier No. HY13781100MG
Pemetrexed disodium hemipenta hydrate is a CNS-penetrant antifolate. Its pentaglutamate form inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT). This product is intended for research use only.
  • CNS-penetrant antifolate
  • Inhibits thymidylate synthase (TS)
  • Inhibits dihydrofolate reductase (DHFR)
  • Inhibits glycinamide ribonucleotide formyltransferase (GARFT)
  • Antitumor activity through inhibition of multiple key folate-requiring enzymes
  • Substrate for the enzyme FPGS
  • Soluble in water