Learn More
Medchemexpress LLC Pemetrexed disodium heptahydrate | 357166-29-1 | MFCD09029394 | 99.9% | 597.5 g/mol | C20H33N5Na2O13 | 1 ML
SDP
Catalog No. 5000243067
Encompass_Preferred
Pemetrexed disodium heptahydrate is supplied as a ready-to-use 10 mM aqueous solution (1 mL) of the heptahydrate form of pemetrexed for laboratory research. It exhibits antifolate activity by inhibiting thymidylate synthase, dihydrofolate reductase, and GARFT, and is provided at high stated purity for controlled assays.
  • Ready-to-use 10 mM aqueous solution, 1 mL.
  • High stated purity (99.9%).
  • Heptahydrate form of pemetrexed (CAS 357166-29-1).
  • Antifolate activity inhibiting TS, DHFR, and GARFT.
  • Suitable for biochemical and cellular assays requiring defined concentrations.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-43067 Supplier Medchemexpress LLC Supplier No. HY10820B10MM 1ML
Pemetrexed disodium heptahydrate is supplied as a ready-to-use 10 mM aqueous solution (1 mL) of the heptahydrate form of pemetrexed for laboratory research. It exhibits antifolate activity by inhibiting thymidylate synthase, dihydrofolate reductase, and GARFT, and is provided at high stated purity for controlled assays.
  • Ready-to-use 10 mM aqueous solution, 1 mL.
  • High stated purity (99.9%).
  • Heptahydrate form of pemetrexed (CAS 357166-29-1).
  • Antifolate activity inhibiting TS, DHFR, and GARFT.
  • Suitable for biochemical and cellular assays requiring defined concentrations.
  • For research use only; not for human or clinical use.