Learn More
Medchemexpress LLC Pemetrexed (disodium heptahydrate) | 357166-29-1 | MFCD09029394 | 99.9% | 597.5 g/mol | C20H33N5Na2O13 | 50 MG
SDP
Catalog No. 5000243052
Encompass_Preferred
Pemetrexed disodium heptahydrate is the disodium heptahydrate salt of pemetrexed, a multitargeted antifolate that interferes with folate-dependent enzymes required for nucleotide biosynthesis. Supplied for research use, it is intended for biochemical and cell-based studies of antitumor activity and folate metabolism.
  • Multitargeted antifolate activity inhibiting TS, DHFR, and GARFT.
  • Heptahydrate form for defined hydration state and reproducibility.
  • High purity suitable for research (99.9%).
  • Provided in small research quantities for laboratory experiments.
  • Molecular weight approximately 597.5 g/mol for formulation calculations.
Catalog No. 50-002-43052 Supplier Medchemexpress LLC Supplier No. HY10820B50MG
Pemetrexed disodium heptahydrate is the disodium heptahydrate salt of pemetrexed, a multitargeted antifolate that interferes with folate-dependent enzymes required for nucleotide biosynthesis. Supplied for research use, it is intended for biochemical and cell-based studies of antitumor activity and folate metabolism.
  • Multitargeted antifolate activity inhibiting TS, DHFR, and GARFT.
  • Heptahydrate form for defined hydration state and reproducibility.
  • High purity suitable for research (99.9%).
  • Provided in small research quantities for laboratory experiments.
  • Molecular weight approximately 597.5 g/mol for formulation calculations.