Learn More
Selleck Chemical LLC Pemetrexed Disodium Hydrate
SDP
Catalog No. 5000001603
Encompass_Preferred
Pemetrexed Disodium Hydrate (LY-231514) is a novel antifolate and antimetabolite for TS DHFR and GARFT with Ki of 1 3 nM 7 2 nM and 65 nM respectively Pemetrexed Disodium Hydrate stimulates autophagy and apoptosis
Catalog No. 50-000-01603 Supplier Selleck Chemical LLC Supplier No. S77851G
Pemetrexed Disodium Hydrate (LY-231514) is a novel antifolate and antimetabolite for TS DHFR and GARFT with Ki of 1 3 nM 7 2 nM and 65 nM respectively Pemetrexed Disodium Hydrate stimulates autophagy and apoptosis