Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pemetrexed disodium salt 2.5 hydrate, 98%, Thermo Scientific Chemicals

Catalog No. AC456180010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456180010 1 g
AC456180050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456180010 Supplier Thermo Scientific Chemicals Supplier No. 456180010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 357166-30-4 | C20H19N5Na2O6 . 5/2 H2O | 516.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357166-30-4
Molecular Formula C20H19N5Na2O6 . 5/2 H2O
Molecular Weight (g/mol) 516.43
MDL Number MFCD13194950
InChI Key NYDXNILOWQXUOF-CETBVFFHNA-L
Synonym pemetrexed disodium hemipenta hydrate
PubChem CID 131673885
IUPAC Name (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid;molecular hydrogen;sodium;pentahydrate
SMILES [Na+].[Na+].NC1=NC2=C(C(CCC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)=CN2)C(=O)N1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 516.43
Percent Purity 98%
Chemical Name or Material Pemetrexed disodium salt 2.5 hydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Suspected of causing genetic defects. May damage fertility. May damage the unborn child.
Precautionary Statement GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.