Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pemetrexed Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014020 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4020 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4020 Supplier MilliporeSigma Supelco Supplier No. PHR1596500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Pemetrexed is an anionic anticancer drug with a narrow therapeutic index. As a folate anti metabolite, it acts as an inhibitor of thymidylate synthase. It can act effectively against a broad-spectrum of solid tumors such as non-small-cell lung and breast cancers in clinical trials. It is also effective against both dihydrofolate reductase and glycinamide ribonucleotide formyl transferase.
TRUSTED_SUSTAINABILITY

Specifications

CAS 357166-29-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H19N5 Na2O6 · 7H2 O
Synonym Pemetrexed disodium heptahydrate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 597.48
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.