Learn More
Cayman Chemical Pemigatinib, Purity: ≥98%, CAS: 1513857-77-6, 5 mg
SDP
Catalog No. 502946316
Encompass_Preferred
An FGFR inhibitor (IC50s = 0.4, 0.5, and 1 nM for FGFR1, FGFR2, and FGFR3, respectively); selective for FGFR1, FGFR2, and FGFR3 over FGFR4 and KDR (IC50s = 30 and 70 nM, respectively), as well as over a panel of 56 tyrosine and serine/threonine kinases at 10 µM; reduces phosphorylation of FGFR, ERK1/2, and STAT5 in KG-1a acute myeloid leukemia cells in a concentration-dependent manner; inhibits growth in a panel of cancer cell lines expressing constitutively active FGFR (GI50s = 3-362 nM); reduces tumor growth in KATO III, KG-1, and RT-112 mouse xenograft model at 0.3 mg/kg
Catalog No. 50-294-6316 Supplier Cayman Chemical Supplier No. 332605 MG
An FGFR inhibitor (IC50s = 0.4, 0.5, and 1 nM for FGFR1, FGFR2, and FGFR3, respectively); selective for FGFR1, FGFR2, and FGFR3 over FGFR4 and KDR (IC50s = 30 and 70 nM, respectively), as well as over a panel of 56 tyrosine and serine/threonine kinases at 10 µM; reduces phosphorylation of FGFR, ERK1/2, and STAT5 in KG-1a acute myeloid leukemia cells in a concentration-dependent manner; inhibits growth in a panel of cancer cell lines expressing constitutively active FGFR (GI50s = 3-362 nM); reduces tumor growth in KATO III, KG-1, and RT-112 mouse xenograft model at 0.3 mg/kg