Learn More
Medchemexpress LLC Pemirolast potassium | 100299-08-9 | 99.99% | 1 ML
SDP
Catalog No. 5000407584
Encompass_Preferred
Pemirolast potassium is an orally active antiallergic agent used for research into bronchial asthma and conjunctivitis. It has been shown to attenuate hypersensitivity reactions, inhibit the release of various mediators from eosinophils and mast cells, and prevent the activation of human eosinophils, thereby alleviating allergic diseases.
  • Orally active antiallergic agent
  • Attenuates hypersensitivity reactions
  • Used for bronchial asthma and conjunctivitis research
  • Inhibits mediator release from eosinophils
  • Prevents activation of human eosinophils
  • Inhibits activation of signal transduction phospholipases in mast cells
  • Reduces cisplatin-induced kaolin intake
Catalog No. 50-004-07584 Supplier Medchemexpress LLC Supplier No. HYB0538A10MM/1ML
Add to Cart
Add to Cart
Pemirolast potassium is an orally active antiallergic agent used for research into bronchial asthma and conjunctivitis. It has been shown to attenuate hypersensitivity reactions, inhibit the release of various mediators from eosinophils and mast cells, and prevent the activation of human eosinophils, thereby alleviating allergic diseases.
  • Orally active antiallergic agent
  • Attenuates hypersensitivity reactions
  • Used for bronchial asthma and conjunctivitis research
  • Inhibits mediator release from eosinophils
  • Prevents activation of human eosinophils
  • Inhibits activation of signal transduction phospholipases in mast cells
  • Reduces cisplatin-induced kaolin intake