Learn More
Medchemexpress LLC Pemirolast potassium | 100299-08-9 | 99.99% | 5 MG
SDP
Catalog No. 5000407583
Encompass_Preferred
Pemirolast potassium is an orally active antiallergic agent effective in research for bronchial asthma and conjunctivitis. This compound acts as an H1 receptor inhibitor, preventing the activation of human eosinophils to inhibit granule protein release, thereby alleviating allergic diseases. It also attenuates hypersensitivity reactions by inhibiting the release of sensory neuropeptides.
  • Orally active antiallergic agent
  • Attenuates hypersensitivity reactions
  • Useful for bronchial asthma and conjunctivitis research
  • Acts as an H1 receptor inhibitor
  • Inhibits eosinophil activation and granule protein release
  • Suppresses degranulation reactions in mast cells
Catalog No. 50-004-07583 Supplier Medchemexpress LLC Supplier No. HYB0538A5MG
Pemirolast potassium is an orally active antiallergic agent effective in research for bronchial asthma and conjunctivitis. This compound acts as an H1 receptor inhibitor, preventing the activation of human eosinophils to inhibit granule protein release, thereby alleviating allergic diseases. It also attenuates hypersensitivity reactions by inhibiting the release of sensory neuropeptides.
  • Orally active antiallergic agent
  • Attenuates hypersensitivity reactions
  • Useful for bronchial asthma and conjunctivitis research
  • Acts as an H1 receptor inhibitor
  • Inhibits eosinophil activation and granule protein release
  • Suppresses degranulation reactions in mast cells