Learn More
Medchemexpress LLC Pemirolast potassium | 100299-08-9 | 99.99% | 50 MG
SDP
Catalog No. 5000408007
Encompass_Preferred
Pemirolast potassium is an orally active antiallergic agent that attenuates Paclitaxel hypersensitivity reactions. It can be used for bronchial asthma and conjunctivitis research.
  • Orally active antiallergic agent
  • Attenuates paclitaxel hypersensitivity reactions
  • Used for bronchial asthma and conjunctivitis research
  • Inhibits A23187-induced LTC4 and ECP release from eosinophils
  • Inhibits PAF-induced and FMLP-induced ECP release from eosinophils
  • Reduces cisplatin-induced kaolin intake and inhibits cisplatin-induced substance P release in the cerebrospinal fluid
Catalog No. 50-004-08007 Supplier Medchemexpress LLC Supplier No. HYB0538A50MG
Pemirolast potassium is an orally active antiallergic agent that attenuates Paclitaxel hypersensitivity reactions. It can be used for bronchial asthma and conjunctivitis research.
  • Orally active antiallergic agent
  • Attenuates paclitaxel hypersensitivity reactions
  • Used for bronchial asthma and conjunctivitis research
  • Inhibits A23187-induced LTC4 and ECP release from eosinophils
  • Inhibits PAF-induced and FMLP-induced ECP release from eosinophils
  • Reduces cisplatin-induced kaolin intake and inhibits cisplatin-induced substance P release in the cerebrospinal fluid