Learn More
Medchemexpress LLC Pemirolast potassium | 100299-08-9 | 99.99% | 500 MG
SDP
Catalog No. 5000408458
Encompass_Preferred
Pemirolast potassium is an orally active antiallergic agent that attenuates hypersensitivity reactions. It can be used for bronchial asthma and conjunctivitis research.
  • Orally active antiallergic agent
  • Attenuates paclitaxel hypersensitivity reactions
  • Used for bronchial asthma research
  • Used for conjunctivitis research
Catalog No. 50-004-08458 Supplier Medchemexpress LLC Supplier No. HYB0538A500MG
Pemirolast potassium is an orally active antiallergic agent that attenuates hypersensitivity reactions. It can be used for bronchial asthma and conjunctivitis research.
  • Orally active antiallergic agent
  • Attenuates paclitaxel hypersensitivity reactions
  • Used for bronchial asthma research
  • Used for conjunctivitis research