Learn More
Cayman Chemical PemIrolastpotasIum salt 100mg
SDP
Catalog No. 502965319
Encompass_Preferred
A histamine H1 receptor antagonist and a mast cell stabilizer that prevents the release of histamine; also blocks the release of substance P, neurokinin A, and calcitonin gene-related peptide from sensory nerves
Catalog No. 50-296-5319 Supplier Cayman Chemical Supplier No. 17889100 MG
A histamine H1 receptor antagonist and a mast cell stabilizer that prevents the release of histamine; also blocks the release of substance P, neurokinin A, and calcitonin gene-related peptide from sensory nerves