Learn More
Cayman Chemical Pemirolast (potassium salt), Purity: ≥98%, CAS: 100299-08-9, 10 mg
SDP
Catalog No. 502968460
Encompass_Preferred
A histamine H1 receptor antagonist and a mast cell stabilizer that prevents the release of histamine; also blocks the release of substance P, neurokinin A, and calcitonin gene-related peptide from sensory nerves
Catalog No. 50-296-8460 Supplier Cayman Chemical Supplier No. 1788910 MG
A histamine H1 receptor antagonist and a mast cell stabilizer that prevents the release of histamine; also blocks the release of substance P, neurokinin A, and calcitonin gene-related peptide from sensory nerves