Learn More
Aviva Systems Biology Corp PEMT Antibody - C-terminal region (ARP43558_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43558_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PEMT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PEMT. Alternate names: MGC2483, PEAMT, PEMPT, PEMT2, PNMTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58567