Learn More
Medchemexpress LLC Pemvidutide (TFA) | 2538014-94-5 | 98.1% | C182H275N39O54.xC2HF3O2 | 25 MG
SDP
Catalog No. 5000356647
Encompass_Preferred
Pemvidutide TFA (ALT-801) is a GLP-1R/GCGR dual agonist that causes significant reductions in body weight, liver fat, and serum lipids. It is used in non-alcoholic steatohepatitis (NASH) and obesity research.
  • Causes striking reductions in body weight, liver fat, and serum lipids
  • Applicable for research in non-alcoholic steatohepatitis (NASH) and obesity
  • Improved NASH outcomes in C57BL6/J mice
  • Lowered hepatic lipids
  • Lowered liver weight
  • Reduced biomarkers of liver injury (plasma ALT and AST)
  • Decreased the inflammation marker galactin-3
  • Reduced the fibrosis marker Col1a1
  • Lowered the NAS score
Catalog No. 50-003-56647 Supplier Medchemexpress LLC Supplier No. 25MGHY145632A
Pemvidutide TFA (ALT-801) is a GLP-1R/GCGR dual agonist that causes significant reductions in body weight, liver fat, and serum lipids. It is used in non-alcoholic steatohepatitis (NASH) and obesity research.
  • Causes striking reductions in body weight, liver fat, and serum lipids
  • Applicable for research in non-alcoholic steatohepatitis (NASH) and obesity
  • Improved NASH outcomes in C57BL6/J mice
  • Lowered hepatic lipids
  • Lowered liver weight
  • Reduced biomarkers of liver injury (plasma ALT and AST)
  • Decreased the inflammation marker galactin-3
  • Reduced the fibrosis marker Col1a1
  • Lowered the NAS score