Learn More
Medchemexpress LLC Pemvidutide (TFA) | C182H275N39O54.xC2HF3O2 | 1 MG
SDP
Catalog No. 5000355887
Encompass_Preferred
Pemvidutide (ALT-801) TFA is a GLP-1R/GCGR dual agonist that demonstrates significant reductions in body weight, liver fat, and serum lipids. It is utilized in research for non-alcoholic steatohepatitis (NASH) and obesity.
  • GLP-1R/GCGR dual agonist
  • Causes significant reductions in body weight
  • Reduces liver fat
  • Reduces serum lipids
  • Used in research for non-alcoholic steatohepatitis (NASH)
  • Used in obesity research
Catalog No. 50-003-55887 Supplier Medchemexpress LLC Supplier No. HY145632A1MG
Pemvidutide (ALT-801) TFA is a GLP-1R/GCGR dual agonist that demonstrates significant reductions in body weight, liver fat, and serum lipids. It is utilized in research for non-alcoholic steatohepatitis (NASH) and obesity.
  • GLP-1R/GCGR dual agonist
  • Causes significant reductions in body weight
  • Reduces liver fat
  • Reduces serum lipids
  • Used in research for non-alcoholic steatohepatitis (NASH)
  • Used in obesity research