Learn More
Medchemexpress LLC Pemvidutide trifluoroacetate | 98.1% | 3873.35 | C182H275N39O54 | 5 MG
SDP
Catalog No. 5000221496
Encompass_Preferred
Pemvidutide TFA is a synthetic peptide acting as a dual agonist of the GLP-1 and glucagon receptors. Supplied for research use, it has been investigated for reductions in body weight, liver fat, and serum lipids, and is intended for preclinical and in-vitro studies only.
  • Dual GLP-1 and glucagon receptor agonist activity.
  • High purity (98.1%).
  • Molecular weight 3873.35 Da.
  • Provided as the trifluoroacetate salt for peptide stability.
  • Available in small milligram sizes for research workflows.
  • Store powder at -80°C (up to 2 years) or -20°C (up to 1 year).
Catalog No. 50-002-21496 Supplier Medchemexpress LLC Supplier No. HY145632A5MG
Pemvidutide TFA is a synthetic peptide acting as a dual agonist of the GLP-1 and glucagon receptors. Supplied for research use, it has been investigated for reductions in body weight, liver fat, and serum lipids, and is intended for preclinical and in-vitro studies only.
  • Dual GLP-1 and glucagon receptor agonist activity.
  • High purity (98.1%).
  • Molecular weight 3873.35 Da.
  • Provided as the trifluoroacetate salt for peptide stability.
  • Available in small milligram sizes for research workflows.
  • Store powder at -80°C (up to 2 years) or -20°C (up to 1 year).