Learn More
Ted Pella Inc PEN FOR FROZEN SECTIONS
SDP
Catalog No. NC9720459
Encompass
PAP Pen for Frozen Sections, Adhesive, Will prevent sections from falling off or wrinkling. Effective with PAP/PAAP, ABC, LAB-SA and immunofluorescent methods. Stable up to 110degreeC, Applications: Immunohistochemical, Microbiology, Histol
Catalog No. NC9720459 Supplier Ted Pella Inc Supplier No. 22302
PAP Pen for Frozen Sections, Adhesive, Will prevent sections from falling off or wrinkling. Effective with PAP/PAAP, ABC, LAB-SA and immunofluorescent methods. Stable up to 110degreeC, Applications: Immunohistochemical, Microbiology, Histol