Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sol-M™ Pen Needle

Catalog No. 22024322
Change view
Click to view available options
Needle Gauge:
31 G
32 G
33 G
Length (Metric) Needle:
4 mm
5 mm
6 mm
8 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (Metric) Needle
22-024-322 33 G 4 mm
22-024-318 31 G 5 mm
22-024-319 31 G 6 mm
22-024-320 31 G 8 mm
22-024-321 32 G 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-024-322 Supplier Sol-M™ Supplier No. PN33532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to provide accurate and reliable insulin delivery. They are engineered for comfortable medication delivery with silicone oil lubrication, sharp triple bevel, and thin wall technology.

  • Double protective cap
  • Tri-bevel anti-coring needle to minimize discomfort
  • Thin-wall needle for optimal medication flow (31G-33G)
  • Compatible with most injector pens

Specifications

Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge 33 G
Latex Free Yes
Length (English) 5/32 in.
Product Type Pen Needle
Disposable Single-use
Length (Metric) Needle 4 mm
Needle Point Style Triple-bevel, Anti-coring
For Use With (Equipment) Insulin
Connection Type Removable
Graduated No
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.