Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pen-Strep (10,000 IU/mL, 10 mg/mL), MP Biomedicals™

Catalog No. p-7236577 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1670249 100 mL
09-167-0246 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN1670249 Supplier MP Biomedicals, Inc Supplier No. 091670249
Only null left
Add to Cart
Add to Cart

Pen-Strep (10,000 IU/mL, 10 mg/mL) is a double antibiotic solution containing 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.

  • The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
  • This solution contains 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.
TRUSTED_SUSTAINABILITY

Specifications

Type Penicillin/Streptomycin
Chemical Name or Material Penicillin-Streptomycin
Quantity 100 mL
Synonym 200X Pen-Strep
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.