Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Glutamine-Penicillin-Streptomycin solution with 200 mM L-glutamine, 10,000 U penicillin and 10 mg steptomycin/mL in 0.9% NaCl, 0.1 mum filtered, BioReagent, suitable for cell culture | MFCD00283636 | 100ML
Catalog No. NC2061741
Encompass
L-Glutamine-Penicillin-Streptomycin solution with 200 mM L-glutamine, 10,000 U penicillin and 10 mg steptomycin/mL in 0.9% NaCl, 0.1 mum filtered, BioReagent, suitable for cell culture | MFCD00283636 | 100ML
Catalog No. NC2061741 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G1146100ML
L-Glutamine-Penicillin-Streptomycin solution with 200 mM L-glutamine, 10,000 U penicillin and 10 mg steptomycin/mL in 0.9% NaCl, 0.1 mum filtered, BioReagent, suitable for cell culture | MFCD00283636 | 100ML